missing translation for 'onlineSavingsMsg'
Learn More

Humanin Antibody, Novus Biologicals™

Product Code. 18048915 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18048915 0.025 mL 25µL
18715993 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18048915 Supplier Novus Biologicals Supplier No. NB10056877SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Humanin Polyclonal specifically detects Humanin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Humanin
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:100-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported by customer review, Immunohistochemistry-Paraffin 1:100
Formulation Whole antisera with 50% Glycerol with 0.025% Sodium Azide
Gene Accession No. Q8IVG9
Gene Alias HN, HUNIN
Gene Symbols MTRNR2L1
Host Species Rabbit
Immunogen The full length peptide sequence from the human MT-RNR2 protein (accesion number Q8IVG9).
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100462977
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.