missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZRANB2 (aa 172-204) Control Fragment Recombinant Protein

Product Code. 30194322
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30194322

Brand: Invitrogen™ RP107114

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84523 (PA5-84523. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZNF265 (Zinc finger protein 265), also known as ZRANB2 (Zinc finger Ran-binding domain-containing protein 2), ZIS, ZIS1 or ZIS2, is a 330 amino acid protein that belongs to the ZRANB2 family. Localized to the nucleus, ZNF265 functions as a splicing factor that is responsible for alternatively splicing Tra-2 beta (transformer-2 beta) transcripts and is thought to interfere with constitutive 5'-splice selection. ZNF265 contains two RanBP2-type zinc fingers through which it conveys its RNA-binding activity. Two isoforms, designated ZIS-1 and ZIS-2, are expressed due to alternative splicing events. Upon DNA damage, ZIS-2 may be phosphorylated by ATM or ATR.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95218
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9406
Name Human ZRANB2 (aa 172-204) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI227013; DKFZp686J1831; DKFZp686N09117; wu:fa96h03; Zfp265; zgc:56597; zinc finger protein 265; Zinc finger Ran-binding domain-containing protein 2; Zinc finger Ran-binding domain-containing protein 2-like protein; zinc finger RANBP2-type containing 2; zinc finger, RAN-binding domain containing 2; zinc finger, splicing; zinc-finger protein 265; zinc-finger, splicing; ZIS; ZIS1; ZIS2; Znf265; ZRANB2
Common Name ZRANB2
Gene Symbol Zranb2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DEDEDDADLSKYNLDASEEEDSNKKKSNRRSRS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.