missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNRF2 (aa 105-182) Control Fragment Recombinant Protein

Product Code. 30197094
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197094

Brand: Invitrogen™ RP108596

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84890 (PA5-84890. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). ZNRF2 (zinc and ring finger 2), also known as RNF202, is a 242 amino acid peripheral membrane protein that contains one RING-type zinc finger and localizes to the lysosome, as well as the endosome and the cell junction. Expressed at high levels in brain tissue, ZNRF2 is thought to function as an E3 ubiquitin-protein ligase that may be involved in the establishment and maintenance of neuronal transmission and plasticity. Upon DNA damage, ZNRF2 is subject to phosphorylation, probably by ATR or ATM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NHG8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 223082
Name Human ZNRF2 (aa 105-182) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1190002C14Rik; AU044205; D6Ertd365e; E3 ubiquitin-protein ligase ZNRF2; protein Ells2; RING finger protein 202; RING-type E3 ubiquitin transferase ZNRF2; RNF202; zinc and ring finger 1; zinc and ring finger 2; zinc and ring finger 2, E3 ubiquitin protein ligase; zinc finger/RING finger 2; zinc/RING finger protein 2; Znrf1; Znrf2
Common Name ZNRF2
Gene Symbol Znrf2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NSSSGPYGSQDSVHSSPEDGGGGRDRPVGGSPGGPRLVIGSLPAHLSPHMFGGFKCPVCSKFVSSDEMDLHLVMCLTK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.