missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNHIT6 (aa 173-275) Control Fragment Recombinant Protein

Product Code. 30209713
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209713

Brand: Invitrogen™ RP91618

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53736 (PA5-53736. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Required for box C/D snoRNAs accumulation involved in snoRNA processing, snoRNA transport to the nucleolus and ribosome biogenesis. [UniProt]
TRUSTED_SUSTAINABILITY

Specifikationer

Accession Number Q9NWK9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54680
Name Human ZNHIT6 (aa 173-275) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2410019A14Rik; BCD1; box C/D snoRNA essential 1 homolog; box C/D snoRNA protein 1; box C/d snoRNA protein 1-like protein; C1orf181; C3H1ORF181; FLJ20729; NY-BR-75; RGD1308723; Serologically defined breast cancer antigen NY-BR-75; zinc finger HIT domain-containing protein 6; zinc finger HIT-type containing 6; zinc finger, HIT type 6; zinc finger, HIT-type containing 6; Znhit6
Common Name ZNHIT6
Gene Symbol ZNHIT6
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KEELMHGECVKEEKDFLKKEIVDDTKVKEEPPINHPVGCKRKLAMSRCETCGTEEAKYRCPRCMRYSCSLPCVKKHKAELTCNGVRDKTAYISIQQFTEMNLL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.