missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZNF80 (aa 248-273) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (42%), Rat (42%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64225 (PA5-64225. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ZNF80 may be involved in transcriptional regulation.
Specifications
Specifications
| Accession Number | P51504 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 7634 |
| Name | Human ZNF80 (aa 248-273) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | pT17; zinc finger protein 80; ZNF80; ZNFpT17 |
| Common Name | ZNF80 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction