missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZNF608 (aa 967-1042) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ZNF608 gene ontology annotations related to this gene include nucleic acid binding.
Specifications
Specifications
| Accession Number | Q9ULD9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 57507 |
| Name | Human ZNF608 (aa 967-1042) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4932417D18Rik; D430007A19Rik; Kiaa1281; LOW QUALITY PROTEIN: zinc finger protein 608; NY-REN-36; renal carcinoma antigen NY-REN-36; Zfp608; Zinc finger protein 608; Znf608 |
| Common Name | ZNF608 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction