missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF498 (aa 208-274) Control Fragment Recombinant Protein

Product Code. 30201127
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30201127 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30201127 Supplier Invitrogen™ Supplier No. RP101593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63411 (PA5-63411. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. As a member of the Krüppel C2H2-type zinc-finger protein family, ZNF498 (zinc finger protein 498), also known as Zinc finger and SCAN domain-containing protein 25 (ZSCAN25), is a 544 amino acid protein. Localized to the nucleus, ZNF498 contains seven C2H2-type zinc fingers and one SCAN box domain. Due to the presence of these domains, ZNF498 is believed to be involved in transcriptional regulation. ZNF498 exists as 4 isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6NSZ9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 221785
Name Human ZNF498 (aa 208-274) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias EG666311; Zfp498; zinc finger and SCAN domain containing 25; zinc finger and SCAN domain-containing protein 25; Zinc finger protein 498; Znf498; Zscan25
Common Name ZNF498
Gene Symbol ZSCAN25
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PAVNPRDQEMAAGFFTAGSQGLGPFKDMALAFPEEEWRHVTPAQIDCFGEYVEPQDCRVSPGGGSKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.