missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF423 (aa 347-407) Control Fragment Recombinant Protein

Product Code. 30208787
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208787

Brand: Invitrogen™ RP107457

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66798 (PA5-66798. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Transcription factor that can both act as an activator or a repressor depending on the context. Plays a central role in BMP signaling and olfactory neurogenesis. Associates with SMADs in response to BMP2 leading to activate transcription of BMP target genes. Acts as a transcriptional repressor via its interaction with EBF1, a transcription factor involved in terminal olfactory receptor neurons differentiation; this interaction preventing EBF1 to bind DNA and activate olfactory-specific genes. Involved in olfactory neurogenesis by participating in a developmental switch that regulates the transition from differentiation to maturation in olfactory receptor neurons. Controls proliferation and differentiation of neural precursors in cerebellar vermis formation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q2M1K9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23090
Name Human ZNF423 (aa 347-407) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias early B-cell factor associated zinc finger protein; Ebfaz; hOAZ; JBTS19; KIAA0760; NPHP14; OAZ; OLF-1/EBF associated zinc finger; olf1/EBF-associated zinc finger protein; Roaz; Smad- and Olf-interacting zinc finger protein; Zfp104; ZFP423; zinc finger protein 423; ZNF423
Common Name ZNF423
Gene Symbol ZNF423
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QPDSSNHSVSPDPVLGSVASMSSATPDSSASVERGSTPDSTLKPLRGQKKMRDDGQGWTKV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.