missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZNF38 (aa 253-401) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54834 (PA5-54834. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ZSCAN21 belongs to the krueppel C2H2-type zinc-finger protein family and is a strong transcriptional activator.
Specifications
Specifications
| Accession Number | Q9Y5A6 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 7589 |
| Name | Human ZNF38 (aa 253-401) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AI326272; CTfin51; hypothetical protein LOC527103; NY-REN-21; renal carcinoma antigen NY-REN-21; RU49; Transcription factor RU49; Zfp38; zfp-38; zinc finger and SCAN domain containing 21; zinc finger and SCAN domain-containing protein 21; zinc finger and SCAN domain-containing protein 21; zinc finger protein 3; zinc finger protein with KRAB and SCAN domains 1; zinc finger proliferation 1; zinc finger protein 38; zinc finger protein 38 (KOX 25); zinc finger protein 38 homolog; zinc finger protein NY-REN-21 antigen; zinc finger with KRAB and SCAN domains 1; Zipro1; ZKSCAN1; ZNF38; ZSCAN21 |
| Common Name | ZNF38 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction