missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF322A (aa 377-402) Control Fragment Recombinant Protein

Product Code. 30180773
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30180773

Brand: Invitrogen™ RP99501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (58%), Rat (58%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61284 (PA5-61284. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ZNF322 contains four exons and spans 23.2kb in chromosome 6p22.1 region, and transcribes a 2.7kb mRNA that encodes a protein with 402 amino acid residues.Through northern blot analysis, ZNF322 was shown to be expressed in every human tissue examined at adult stage and during embryonic developmental stages from 80 days to 24 weeks. When ZNF322 was overexpressed in COS-7 cells, ZNF322-EGFP fusion protein is detected in the nucleus and cytoplasm. Reporter gene assays show that ZNF322 is a transcriptional activator. Furthermore, overexpression of ZNF322 in COS-7 cells activates the transcriptional activity of SRE and AP-1. Together, these results suggest that ZNF322 is a member of the zinc-finger transcription factor family and may act as a positive regulator in gene transcription mediated by the MAPK signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6U7Q0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79692
Name Human ZNF322A (aa 377-402) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias HCG12; HLA complex group 12; Zfp322a; Zinc finger protein 322; zinc finger protein 322 A; Zinc finger protein 388; zinc finger protein 489; ZNF322; ZNF322A; ZNF388; ZNF489
Common Name ZNF322A
Gene Symbol ZNF322
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KPFVCNVSEKGLELSPPHASEASQMS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.