missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZNF318 (aa 186-324) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55489 (PA5-55489. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ZNF318 encodes a nuclear protein with a zinc finger motif of the Cys2-His2 type that is a novel corepressor of androgen receptor (AR).
Specifications
Specifications
| Accession Number | Q5VUA4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 24149 |
| Name | Human ZNF318 (aa 186-324) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2610034E08Rik; D530032D06Rik; endocrine regulator; endocrine regulatory protein; HRIHFB2436; testicular zinc finger; testicular zinc finger protein; Tzf; ZFP318; Zinc finger protein 318; Znf318 |
| Common Name | ZNF318 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction