missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF306 (aa 13-45) Control Fragment Recombinant Protein

Product Code. 30212439
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212439

Brand: Invitrogen™ RP108441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84442 (PA5-84442. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ZNF306 is a protein coding gene. Transcriptional factor that binds to the consensus sequence 5'-[GT][AG][AGT]GGGG-3' and acts as a repressor of autophagy. Specifically represses expression of genes involved in autophagy and lysosome biogenesis/function such as MAP1LC3B, ULK1 or WIPI2. Associates with chromatin at the ITGB4 and VEGF promoters. Also acts as a transcription activator and promotes cancer cell progression and/or migration in various tumors and myelomas.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BRR0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 80317
Name Human ZNF306 (aa 13-45) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2810435N07Rik; AA408594; AI132359; dJ874C20.1; dJ874C20.1.; mszf35; SCAN-KRAB-zinc finger gene 1; SCAN-KRAB-zinc finger protein; Skz1; Zf47; ZFP306; Zfp307; Zfp47; zfp-47; Zinc finger and SCAN domain-containing protein 13; zinc finger protein 306; Zinc finger protein 307; zinc finger protein 309; Zinc finger protein 47 homolog; zinc finger protein with KRAB and SCAN domains 3; zinc finger protein zfp47; zinc finger with KRAB and SCAN domains 3; Zkscan3; ZNF306; ZNF309; ZSCAN13; ZSCAN35
Common Name ZNF306
Gene Symbol ZKSCAN3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AQSTEDQMELLVIKVEEEEAGFPSSPDLGSEGS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.