missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZNF295 (aa 998-1066) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ZNF295 contains 1 BTB (POZ) domain and 8 C2H2-type zinc fingers. ZNF295 may be involved in transcriptional regulation.
Specifications
Specifications
| Accession Number | Q9ULJ3 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 49854 |
| Name | Human ZNF295 (aa 998-1066) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 5430437K12Rik; B430213I24Rik; KIAA1227; mKIAA1227; ZBTB21; Zfp295; zinc finger and BTB domain containing 21; zinc finger and BTB domain-containing protein 21; Zinc finger protein 295; ZNF295 |
| Common Name | ZNF295 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction