missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ZNF206 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4020
This item is not returnable.
View return policy
Description
ZNF206 Polyclonal specifically detects ZNF206 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| ZNF206 | |
| Polyclonal | |
| Unconjugated | |
| Q96SZ4 | |
| ZSCAN10 | |
| E. coli-derived recombinant human ZNF206 Met1-Val283 Accession # Q96SZ4 | |
| 100 μg | |
| Primary | |
| Detects human ZNF206 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human ZNF24 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Zfp206, ZSCAN10, ZSCAN10 zinc finger and SCAN domain containing 10 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 84891 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction