missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF197 (aa 253-325) Control Fragment Recombinant Protein

Product Code. 30195711
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195711

Brand: Invitrogen™ RP101574

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (27%), Rat (27%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84210 (PA5-84210. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ZNF197 product belongs to the zinc finger protein superfamily, members of which are regulatory proteins characterized by nucleic acid-binding zinc finger domains. The encoded protein contains 20 tandemly arrayed C2H2-type zinc fingers, a Kruppel-associated box (KRAB) domain, and a SCAN box. This transcript turns over rapidly and contains 3' UTR AUUUA motifs, which are often a hallmark of rapid turnover. It is overexpressed in some thyroid papillary carcinomas. ZNF197 is located in a cluster of zinc finger genes at 3p21. Two alternatively spliced transcripts encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14709
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10168
Name Human ZNF197 (aa 253-325) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias D3S1363E; P18; pVHL-associated KRAB domain-containing protein; VHLaK; VHL-associated KRAB-A domain-containing protein; zinc finger protein 166; zinc finger protein 197; Zinc finger protein 197-like protein; zinc finger protein 20; zinc finger protein with KRAB and SCAN domains 9; ZKSCAN9; ZNF166; ZNF197; ZnF20; ZSCAN41
Common Name ZNF197
Gene Symbol ZNF197
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VTSLEWETMTENEEVTSKPSSSQRADSHKGTSKRLQGSVPQVLDFEEECEWQVLASQWGNETDERADTVKKVS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.