missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Human ZNF179 (aa 427-540) Control Fragment Recombinant Protein
Klicka för att se tillgängliga alternativ
Quantity:
100 μL
Förpackningsstorlek:
100µL
Beskrivning
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53402 (PA5-53402. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The specific function of this protein remains unknown. This gene encodes a member of the RING finger protein family of transcription factors. The protein is primarily expressed in brain. The gene is located within the Smith-Magenis syndrome region on chromosome 17.
Specifikationer
Specifikationer
| Accession Number | Q9ULX5 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 7732 |
| Name | Human ZNF179 (aa 427-540) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AI854096; Bfb; bfp; Brain finger protein; LOW QUALITY PROTEIN: RING finger protein 112; neurolastin; R75574; RING finger protein 112; Rnf112; Zfp179; zinc finger protein 179; ZNF179 |
| Common Name | ZNF179 |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering