missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ZNF143 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6804-SP
This item is not returnable.
View return policy
Description
ZNF143 Monoclonal specifically detects ZNF143 in Human samples. It is validated for Immunocytochemistry.
Specifications
| ZNF143 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| P52747 | |
| ZNF143 | |
| E. coli-derived recombinant human ZNF143 Gly459-Asp638 Accession # P52747 | |
| 25 μg | |
| Primary | |
| Detects human ZNF143 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ZNF24, rhZNF206, rhZNF281, rhDC-SCRIPT/ZNF366, rhZNF423, or recombinant mouse Bcl-6 is observed. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 677314 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hStaf, pHZ-1, SBF, SBFzinc finger protein 143 (clone pHZ-1), Selenocysteine tRNA gene transcription-activating factor, SPH-binding factor, STAF, STAFpHZ-1, transcriptional activator Staf, zinc finger protein 143 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7702 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction