missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZIP14 (aa 89-154) Control Fragment Recombinant Protein

Product Code. 30209514
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209514

Brand: Invitrogen™ RP108930

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140224 (PA5-140224. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The zinc transporter ZIP14, also known as SLC39A14, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP14 is a glycosylated multipass plasma membrane protein that belongs to the ZIP transporter subfamily LIV-1. ZIP14 has been shown to contribute to the hypozincemia of inflammation and infection and is regulated in the liver by IL-6. In addition to zinc, ZIP14 is also involved in the cellular uptake of non-transferrin-bound iron as well as iron bound to transferrin.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15043
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23516
Name Human ZIP14 (aa 89-154) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias cig19; Factor for adipocyte differentiation 123; Fad123; FAD-123; HMNDYT2; Kiaa0062; LIV-1 subfamily of ZIP zinc transporter 4; LZT-Hs4; NET34; SLC39A14; solute carrier family 39 (metal ion transporter), member 14; solute carrier family 39 (zinc transporter), member 14; solute carrier family 39 member 14; Zinc transporter ZIP14; Zip14; ZIP-14; Zrt- and Cig19; Zrt- and Irt-like protein 14; Zrt-, Irt-like protein 14
Common Name ZIP14
Gene Symbol SLC39A14
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence STCFSSGDLFTAHNFSEQSRIGSSELQEFCPTILQQLDSRACTSENQENEENEQTEEGRPSAVEVW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.