missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZIP10 (aa 69-148) Control Fragment Recombinant Protein

Product Code. 30202481
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202481

Brand: Invitrogen™ RP107478

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ZIP10, also known as Slc39A10, is a widely expressed zinc transporter with nine transmembrane domains. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP10 mRNA was found to be significantly decreased in the intestines and kidneys of hypothyroid rats and increased in those of hyperthyroid rats, indicating that ZIP10 is positively regulated by thyroid hormones. ZIP10 mRNA was also found to be upregulated in invasive and metastatic breast cancer and cell lines, suggesting that ZIP10 could serve as a possible marker for the metastatic phenotype and possibly a target for novel treatment strategies. At least three isoforms of ZIP10 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9ULF5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57181
Name Human ZIP10 (aa 69-148) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2900042E17Rik; 5430433I10; Kiaa1265; LZT-Hs2; mKIAA1265; Slc39a10; solute carrier family 39 (metal ion transporter), member 10; solute carrier family 39 (zinc transporter), member 10; solute carrier family 39 member 10; Zinc transporter ZIP10; ZIP10; ZIP-10; Zrt- and Irt-like protein 10
Common Name ZIP10
Gene Symbol SLC39A10
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ERYGENGRLSFFGLEKLLTNLGLGERKVVEINHEDLGHDHVSHLDILAVQEGKHFHSHNHQHSHNHLNSENQTVTSVSTK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.