missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZIC4 (aa 7-107) Control Fragment Recombinant Protein

Product Code. 30197178
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197178

Brand: Invitrogen™ RP102566

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56392 (PA5-56392. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development, and have been associated with X-linked visceral heterotaxy and holoprosencephaly type 5. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 1, a related family member on chromosome 3. Heterozygous deletion of these linked genes is involved in Dandy-Walker malformation, which is a congenital cerebellar malformation. Multiple transcript variants have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N9L1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84107
Name Human ZIC4 (aa 7-107) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Zic family member 4; ZIC4; zinc family member 4 protein HZIC4; zinc finger protein of the cerebellum 4; Zinc finger protein ZIC 4
Common Name ZIC4
Gene Symbol ZIC4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LVMRKRLRLYRNTLKESSSSSGHHGPQLTAASSPSVFPGLHEEPPQASPSRPLNGLLRLGLPGDMYARPEPFPPGPAARSDALAAAAALHGYGGMNLTVNL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.