missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZFAND6 (aa 112-140) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (57%), Rat (57%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139818 (PA5-139818. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ZA20D3 is a zinc-finger protein located on chromosome 15.
Specifications
Specifications
| Accession Number | Q6FIF0 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 54469 |
| Name | Human ZFAND6 (aa 112-140) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 3110005P07Rik; AN1-type zinc finger protein 6; associated with PRK1 protein; associated with Prkcl1; Awp1; HT032; protein associated with PRK1; Za20d3; ZFAND5B; ZFAND6; zgc:101121; zinc finger A20 domain-containing protein 3; zinc finger AN1-type containing 6; zinc finger, A20 domain containing 3; zinc finger, AN1-type domain 6 |
| Common Name | ZFAND6 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction