missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ZEB2/SIP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£180.00 - £431.00
Specifications
| Antigen | ZEB2 |
|---|---|
| Clone | 923328 |
| Dilution | Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Immunohistochemistry, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18728418
![]() |
R&D Systems
MAB73782-SP |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18799668
![]() |
R&D Systems
MAB73782 |
100 μg |
£431.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZEB2 Monoclonal specifically detects ZEB2 in Human samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| ZEB2 | |
| Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HSPC082, KIAA0569FLJ42816, SIP1, SIP1HSPC082, Smad-interacting protein 1, SMADIP1, SMADIP1ZFHX1BSIP-1, ZFHX1B, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b | |
| ZEB2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 923328 | |
| Immunohistochemistry, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| O60315 | |
| 9839 | |
| E. coli-derived recombinant human ZEB2/SIP1 Asn363-Lys537 Accession # O60315 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ZEB2/SIP1 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title