missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZEB1 (aa 565-694) Control Fragment Recombinant Protein

Product Code. 30193799
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30193799

Brand: Invitrogen™ RP100723

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82982 (PA5-82982. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ZEB1 (Zinc finger E-box-binding homeobox 1) acts as a transcriptional repressor. It inhibits interleukin-2 (IL-2) gene expression. ZEB1 enhances or represses the promoter activity of the ATP1A1 gene depending on the quantity of cDNA and depending on the cell type. It represses E-cadherin promoters and induces an epithelial-mesenchymal transition (EMT) by recruiting SMARCA4/BRG1. ZEB1 also represses BCL6 transcription in the presence of the corepressor CTBP1. It postively regulates neuronal differentiation and represses RCOR1 transcription activation during neurogenesis. Mutations in the gene can result in corneal dystrophy, posterior polymorphous and corneal dystrophy, Fuchs endothelial 6.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P37275
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6935
Name Human ZEB1 (aa 565-694) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias [delta]EF1; 3110032K11Rik; AREB6; BZP; Delta EF1; delta-crystallin enhancer binding factor 1; deltaEF1; DELTA-EF1; FECD6; LOW QUALITY PROTEIN: zinc finger E-box-binding homeobox 1; MEB1; MGC133261; Negative regulator of IL2; Nil2; NIL2A; NIL-2 A; NIL-2-A; NIL-2-A zinc finger protein; OTTHUMP00000019405; posterior polymorphous corneal dystrophy 3; PPCD3; RP11-472N13.4; Tcf18; TCF8; TCF-8; Transcription factor 8; transcription factor 8 (represses interleukin 2 expression); Tw; twirler; ZEB; Zeb1; Zfhep; Zfh x 1 A; Zf x 1 A; Zf x 1 ha; zinc finger E-box binding homeobox 1; zinc finger E-box-binding homeobox 1; Zinc finger homeobox protein 1 A; zinc finger homeodomain enhancer-binding protein
Common Name ZEB1
Gene Symbol ZEB1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EAEKPESSVSSATGDGNLSPSQPPLKNLLSLLKAYYALNAQPSAEELSKIADSVNLPLDVVKKWFEKMQAGQISVQSSEPSSPEPGKVNIPAKNNDQPQSANANEPQDSTVNLQSPLKMTNSPVLPVGST
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.