missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZC3H12A Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP1040

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15275039

  • £112.00 / 50µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ZC3H12A Synthetic Peptide, PEP-1040, from Invitrogen.

This peptide corresponds to 17 amino acids near the center of human ZC3H12A.PEP-1040 can be used as a blocking peptide with polyclonal antibody PA5-20926.
Specifications

Specifications

80149
purified
Human ZC3H12A
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q5D1E8
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
ZC3H12A
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers