missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ZBP1/DLM-1/DAI Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6309-SP
This item is not returnable.
View return policy
Description
ZBP1/DLM-1/DAI Polyclonal specifically detects ZBP1/DLM-1/DAI in Human samples. It is validated for Western Blot.
Specifications
| ZBP1/DLM-1/DAI | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C20orf183, chromosome 20 open reading frame 183, DAI, DLM1, DLM-1, DLM1dJ718J7.3, DNA-dependent activator of interferon regulatory factors, DNA-dependent activator of IRFs, Tumor stroma and activated macrophage protein DLM-1, ZBP1 Z-DNA binding protein 1, Z-DNA binding protein 1, Z-DNA-binding protein 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 81030 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9H171 | |
| ZBP1 | |
| E. coli-derived recombinant human ZBP1/DLM-1/DAI Ala2-Ile429 Accession # Q9H171 | |
| 25 μg | |
| Primary | |
| Detects human ZBP1/DLM-1/DAI in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction