missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human ZAG Platinum ELISA Kit
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
Includes
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human ZAG
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
| Assay Sensitivity | 0.174ng/mL |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 96 Tests |
| Sample Volume | 10 μL |
| Synonym | Zinc alpha 2-glycoprotein, AZGP1, ZA2G, ZAG |
| Target | ZAG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction