missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human YY1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3784-SP
This item is not returnable.
View return policy
Description
YY1 Polyclonal specifically detects YY1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| YY1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DELTA, Delta transcription factor, INO80SYY-1, NF-E1, NF-E1INO80 complex subunit S, transcriptional repressor protein YY1, UCRBP, Yin and yang 1, Yin and Yang 1 protein, YIN-YANG-1, YY1 transcription factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7528 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P25490 | |
| YY1 | |
| E. coli-derived recombinant human YY1 His75-Gln275 Accession # P25490 | |
| 25 μg | |
| Primary | |
| Detects human YY1 in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction