missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human YLAT2 (aa 1-44) Control Fragment Recombinant Protein

Product Code. 30207013
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30207013 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30207013 Supplier Invitrogen™ Supplier No. RP91213

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (66%), Rat (66%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62233 (PA5-62233. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Involved in the sodium-independent uptake of dibasic amino acids and sodium-dependent uptake of some neutral amino acids. Requires coexpression with SLC3A2/4F2hc to mediate the uptake of arginine, leucine and glutamine. Also acts as an arginine/glutamine exchanger, following an antiport mechanism for amino acid transport, influencing arginine release in exchange for extracellular amino acids. Plays a role in nitric oxide synthesis in human umbilical vein endothelial cells (HUVECs) via transport of L-arginine. Involved in the transport of L-arginine in monocytes. Reduces uptake of ornithine in retinal pigment epithelial (RPE) cells. [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92536
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9057
Name Human YLAT2 (aa 1-44) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI643885; amino acid permease; cationic amino acid transporter, y+ system; KIAA0245; LAT-2; LAT3; Slc7a6; solute carrier family 7 (amino acid transporter light chain, y+L system), member 6; solute carrier family 7 (cationic amino acid transporter, y+ system), member 6; solute carrier family 7 member 6; solute carriersolute carrier family 7 (cationic amino acid transporter, y+ system), member 6; y(+)L-type amino acid transporter 2; Y+L amino acid transporter 2; Y+LAT2; y+LAT-2
Common Name YLAT2
Gene Symbol SLC7A6
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MEAREPGRPTPTYHLVPNTSQSQVEEDVSSPPQRSSETMQLKKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.