missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human XPG Control Fragment Recombinant Protein

Product Code. 30210128
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210128

Brand: Invitrogen™ RP95291

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (43%), Rat (43%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62177 (PA5-62177. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Xeroderma pigmentosum type G (XPG) is a human genetic disease exhibiting extreme sensitivity to sunlight. The XPG protein, a member of the flap endonuclease 1 (FEN-1) structure-specific DNA repair endonuclease family, is an enzyme essential for DNA repair of the major kinds of solar ultraviolet (UV)-induced DNA damages. Human XPG nuclease makes the 3' incision during nucleotide excision repair of DNA. The enzyme cleaves model DNA bubble structures specifically near the junction of unpaired DNA with a duplex region. A 29-amino acid region of human XPG (residues 981-1009) contains the PCNA binding activity. A conserved arginine in XPG (Arg992) is crucial for its PCNA binding activity. Replication Protein A (RPA) binds specifically and directly to two excision repair proteins, the xeroderma pigmentosum damage-recognition protein XPA and the endonuclease XPG.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P28715
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2073
Name Human XPG Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias COFS 3; COFS3; DNA excision repair protein ERCC-5; DNA repair protein complementing XP-G cells; DNA repair protein complementing XP-G cells homolog; ERCC 5; ERCC excision repair 5, endonuclease; ERCC5; ERCC5-201; ERCM 2; ERCM2; excision repair cross-complementation group 5; excision repair cross-complementing rodent repair deficiency, complementation group 5; OTTHUMP00000064902; RP11-484I6.5; UVDR; Xe; xeroderma pigmentosum; Xeroderma pigmentosum group G-complementing protein; xeroderma pigmentosum group G-complementing protein homolog; xeroderma pigmentosum, complementation group G; XPG; XPGC; XPG-complementing protein
Common Name XPG
Gene Symbol ERCC5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EKEFELLDKAKRKTQKRGITNTLEESSSLKRKRLSDSKRKNTCGGFLGETCLSESSDGSSSEDAESSSLMNVQRRTAAKEPKTSASDSQNSVKEAPVK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.