missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human XPE/DDB2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3297
This item is not returnable.
View return policy
Beschreibung
XPE/DDB2 Polyclonal specifically detects XPE/DDB2 in Human samples. It is validated for Western Blot.
Spezifikation
| XPE/DDB2 | |
| Polyclonal | |
| Unconjugated | |
| Q92466 | |
| DDB2 | |
| E. coli-derived recombinant human XPE/DDB2 Met1-Lys427 Accession # Q92466 | |
| 100 μg | |
| Primary | |
| Detects human XPE/DDB2 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| damage-specific DNA binding protein 2 (48kD), damage-specific DNA binding protein 2, 48kDa, Damage-specific DNA-binding protein 2, DDB p48 subunit, DDB2, DDBb, DNA damage-binding protein 2, FLJ34321, UV-damaged DNA-binding protein 2, UV-DDB 2, UV-DDB2, xeroderma pigmentosum group E protein, XP5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1643 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur