missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human XPE/DDB2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3297-SP
This item is not returnable.
View return policy
Description
XPE/DDB2 Polyclonal specifically detects XPE/DDB2 in Human samples. It is validated for Western Blot.
Specifications
| XPE/DDB2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| damage-specific DNA binding protein 2 (48kD), damage-specific DNA binding protein 2, 48kDa, Damage-specific DNA-binding protein 2, DDB p48 subunit, DDB2, DDBb, DNA damage-binding protein 2, FLJ34321, UV-damaged DNA-binding protein 2, UV-DDB 2, UV-DDB2, xeroderma pigmentosum group E protein, XP5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1643 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| Q92466 | |
| DDB2 | |
| E. coli-derived recombinant human XPE/DDB2 Met1-Lys427 Accession # Q92466 | |
| 25 μg | |
| Primary | |
| Detects human XPE/DDB2 in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction