missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human XIAP Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8221
This item is not returnable.
View return policy
Description
XIAP Monoclonal specifically detects XIAP in Human samples. It is validated for Immunohistochemistry, Neutralization.
Specifications
| XIAP | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL, Neutralization 10 ug/mL | |
| P98170 | |
| XIAP | |
| E. coli-derived recombinant human XIAP Met1-Ser497 Accession # P98170 | |
| 100 μg | |
| Primary | |
| Detects human XIAP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) cIAP-1 or rh cIAP-2 is observed. Neutralizes human XIAP and human XIAP-BIR2 bioactivity. Does not neutralize XIAP-BIR3 or Livin bioactivity. | |
| Human | |
| Purified |
| Immunohistochemistry, Neutralization | |
| 117318 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| API3X-linked inhibitor of apoptosis protein, apoptosis inhibitor 3, baculoviral IAP repeat-containing 4, baculoviral IAP repeat-containing protein 4, BIRC4, BIRC4XLP2, E3 ubiquitin-protein ligase XIAP, EC 6.3.2.-, hIAP3, hIAP-3, hILP, IAP3, IAP-3, IAP-like protein, ILP, ILP1, Inhibitor of apoptosis protein 3, MIHA, X-linked IAP, X-linked inhibitor of apoptosis | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 331 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 degreesC as supplied. 3 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction