missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human WWP1 (aa 98-193) Control Fragment Recombinant Protein

Codice prodotto. 30207930
Click to view available options
Quantity:
100 μL
Dimensione della confezione:
100µL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 30207930

Marca: Invitrogen™ RP93779

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54715 (PA5-54715. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 4 tandem WW domains and a HECT (homologous to the E6-associated protein carboxyl terminus) domain. The encoded protein belongs to a family of NEDD4-like proteins, which are E3 ubiquitin-ligase molecules and regulate key trafficking decisions, including targeting of proteins to proteosomes or lysosomes. Alternative splicing of this gene generates at least 6 transcript variants; however, the full length nature of these transcripts has not been defined. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifica

Accession Number Q9H0M0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11059
Name Human WWP1 (aa 98-193) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 8030445B08Rik; AIP5; atrophin-1 interacting protein 5; atrophin-1-interacting protein 5; DKFZp434D2111; HECT-type E3 ubiquitin transferase WWP1; hSDRP1; NEDD4-like E3 ubiquitin-protein ligase WWP1; Nedd-4-like ubiquitin-protein ligase; SDRP1; TGIF-interacting ubiquitin ligase 1; Tiul1; Unknown (protein for MGC:129005); WW domain containing E3 ubiquitin protein ligase 1; WW domain-containing protein 1; Wwp1
Common Name WWP1
Gene Symbol Wwp1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TIDLKQALLIHNRKLERVKEQLKLSLENKNGIAQTGELTVVLDGLVIEQENITNCSSSPTIEIQENGDALHENGEPSARTTARLAVEGTNGIDNHV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato