missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human WNT9A Full-length ORF (NP_003386, 1 a.a. - 365 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16101892
Change view
Click to view available options
Quantity:
2 μg
Unit Size:
2µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16101892 2 μg 2µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16101892 Supplier Abnova™ Supplier No. H00007483P01.2ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The WNT gene family consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is expressed in gastric cancer cell lines. The protein encoded by this gene shows 75% amino acid identity to chicken Wnt14, which has been shown to play a central role in initiating synovial joint formation in the chick limb. This gene is clustered with another family member, WNT3A, in the chromosome 1q42 region. [provided by RefSeq]

Sequence: MLDGSPLARWLAAAFGLTLLLAALRPSAAYFGLTGSEPLTILPLTLEPEAAAQAHYKACDRLKLERKQRRMCRRDPGVAETLVEAVSMSALECQFQFRFERWNCTLEGRYRASLLKRGFKETAFLYAISSAGLTHALAKACSAGRMERCTCDEAPDLENREAWQWGGCGDNLKYSSKFVKEFLGRRSSKDLRARVDFHNNLVGVKVIKAGVETTCKCHGVSGSCTVRTCWRQLAPFHEVGKHLKHKYETALKVGSTTNEAAGEAGAISPPRGRASGAGGSDPLPRTPELVHLDDSPSFCLAGRFSPGTAGRRCHREKNCESICCGRGHNTQSRVVTRPCQCQVRWCCYVECRQCTQREEVYTCKG

Specifications

Accession Number NP_003386
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7483
Molecular Weight (g/mol) 65.78kDa
Name WNT9A (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MLDGSPLARWLAAAFGLTLLLAALRPSAAYFGLTGSEPLTILPLTLEPEAAAQAHYKACDRLKLERKQRRMCRRDPGVAETLVEAVSMSALECQFQFRFERWNCTLEGRYRASLLKRGFKETAFLYAISSAGLTHALAKACSAGRMERCTCDEAPDLENREAWQWGGCGDNLKYSSKFVKEFLGRRSSKDLRARVDFHNNLVGVKVIKAGVETTCKCHGVSGSCTVRTCWRQLAPFHEVGKHLKHKYETALKVGSTTNEAAGEAGAISPPRGRASGAGGSDPLPRTPELVHLDDSPSFCLAGRFSPGTAGRRCHREKNCESICCGRGHNTQSRVVTRPCQCQVRWCCYVECRQCTQREEVYTCKG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC138165/MGC141991/WNT14
Common Name WNT9A
Gene Symbol WNT9A
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.