missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Wnt-9b Recombinant Protein, PeproTech®

Product Code. 17860173
Click to view available options
Quantity:
10 x 100 μg
100 μg
2.5 x 100 μg
20 μg
5 μg
5 x 100 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
Pack of 5
This item is not returnable. View return policy

Product Code. 17860173

Brand: Gibco™ 12049100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

120-49-1MG will be provided as 10 x 100 μg (120-49-100UG). 120-49-250UG will be provided as multiple 50 μg (120-49-50UG) and 100 μg (120-49-100UG) vials. 120-49-500UG will be provided as 5 x 100 μg (120-49-100UG). Recombinant Human Wnt-9b is a monomeric glycoprotein containing 335 amino acid residues and has a calculated molecular weight of 36.9 kDa. Due to glycosylation, Recombinant Human Wnt-9b migrates at an apparent molecular weight of approximately 49-54 kDa by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

WNT9B, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14905
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 7484
Molecular Weight (g/mol) 36.9 kDa
Name Human Wnt-9 b
Quantity 100 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias clf; clf1; protein Wnt-14 b; Protein Wnt-15; Protein Wnt-9 b; UNQ6973/PRO21956; wingless-type MMTV integration site 15; wingless-type MMTV integration site 9 B; wingless-type MMTV integration site family member 9 B; wingless-type MMTV integration site family, member 15; wingless-type MMTV integration site family, member 9 B; Wnt family member 9 B; Wnt14b; wnt-14 b; Wnt15; wnt-15; Wnt9b
Common Name WNT9B
Gene Symbol Wnt9b
Biological Activity Determined by its ability to induce alkaline phosphatase production by CCL-226 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence SYFGLTGREV LTPFPGLGTA AAPAQGGAHL KQCDLLKLSR RQKQLCRREP GLAETLRDAA HLGLLECQFQ FRHERWNCSL EGRMGLLKRG FKETAFLYAV SSAALTHTLA RACSAGRMER CTCDDSPGLE SRQAWQWGVC GDNLKYSTKF LSNFLGSKRG NKDLRARADA HNTHVGIKAV KSGLRTTCKC HGVSGSCAVR TCWKQLSPFR ETGQVLKLRY DSAVKVSSAT NEALGRLELW APARQGSLTK GLAPRSGDLV YMEDSPSFCR PSKYSPGTAG RVCSREASCS SLCCGRGYDT QSRLVAFSCH CQVQWCCYVE CQQCVQEELV YTCKH
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.