missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Wnt-9a Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB31571-SP
This item is not returnable.
View return policy
Description
Wnt-9a Monoclonal specifically detects Wnt-9a in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Wnt-9a | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| O14904.2 | |
| WNT9A | |
| E. coli-derived recombinant human Wnt-9a Pro48-Glu92 & Arg231-Phe304 Accession # O14904.2 | |
| 25 μg | |
| Primary | |
| Detects recombinant human Wnt-9a in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Wnt-2, -7a, -7b, recombinant mouse Wnt-1, -2b, -3a, -4, -5a, -5b, -8a, -8b, -9b, -10a, -10b, -11, or -16 is observed | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 326707 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC138165, MGC141991, Protein Wnt-14, wingless-type MMTV integration site family, member 14, wingless-type MMTV integration site family, member 9A, Wnt-14, WNT14protein Wnt-9a, Wnt9a | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7483 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction