missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Wnt-7a Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3008-100
This item is not returnable.
View return policy
Description
Wnt-7a Monoclonal specifically detects Wnt-7a in Human samples. It is validated for Immunocytochemistry.
Specifications
| Wnt-7a | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| protein Wnt-7a, proto-oncogene Wnt7a protein, wingless-type MMTV integration site family, member 7A, Wnt7a | |
| Chinese hamster ovary cell line CHO-derived recombinant human Wnt-7a Met1-Lys349 Accession # O00755 | |
| 100 μg | |
| Primary | |
| Detects human Wnt-7a in direct ELISAs. Cross-reactivity with Wnt-7b has not been determined. | |
| Human | |
| IgG2b |
| Immunocytochemistry | |
| 946730 | |
| Immunocytochemistry 5-25 ug/mL | |
| O00755 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7476 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction