missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Wnt-16b Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7790-SP
This item is not returnable.
View return policy
Description
Wnt-16b Monoclonal specifically detects Wnt-16b in Human samples. It is validated for Immunocytochemistry.
Specifications
| Wnt-16b | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AHCP, WNT16, Wnt16b | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51384 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Immunocytochemistry | |
| 948309 | |
| Immunocytochemistry 8-25 ug/mL | |
| Q9UBV4 | |
| WNT16 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Wnt-16 Asn30-Lys365 Accession # Q9UBV4 | |
| 25 μg | |
| Primary | |
| Detects human Wnt-16b in ELISA. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction