missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Wnt-10b Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB71963-100
This item is not returnable.
View return policy
Description
Wnt-10b Monoclonal specifically detects Wnt-10b in Human samples. It is validated for ELISA.
Specifications
| Wnt-10b | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| protein Wnt-10b, Protein Wnt-12, SHFM6WNT-10B protein, wingless-type MMTV integration site family, member 10B, Wnt10b, Wnt-12, WNT12 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Wnt-10b, Met1-Lys389, Accession No. O00744 | |
| 100 μg | |
| 7480.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| ELISA | |
| 904903R | |
| ELISA | |
| O00744 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Wnt-10b in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction