missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human WNK2 (aa 1676-1803) Control Fragment Recombinant Protein

Product Code. 30201536
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201536

Brand: Invitrogen™ RP91472

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (60%), Rat (60%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53440 (PA5-53440. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

WNK2 is a serine/threonine kinase involved in signal transduction and cellular communication. WNK kinases contain a lysine upstream of the traditional position, within a glycine string. This lysine functions as an anchor and orients ATP through interactions with the alpha and beta phosphoryl groups. The catalytic domains of WNK2, WNK3 and WNK4 are 95% homologous to WNK1. The human WNK1 gene encodes a 2,382 amino acid protein that is primarily expressed in heart, kidney, muscle and distal nephron. The human WNK3 gene encodes a protein that is primarily expressed in brain; the human WNK4 gene encodes a 1,243 amino acid protein that is expressed in kidney. Aberrant function of WNK kinases and their associated signaling pathways are implicated in hypertension, increased renal salt reabsorption and impaired K+ and H+ excretion.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y3S1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 65268
Name Human WNK2 (aa 1676-1803) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1810073P09Rik; Antigen NY-CO-43; AW122246; ESTM15; Kiaa1760; kinase deficient protein; LOW QUALITY PROTEIN: serine/threonine-protein kinase WNK2; mitogen-activated protein kinase kinase kinase; mKIAA1760; NY-CO-43; P/OKcl0.13; PRKWNK2; protein kinase lysine-deficient 2; Protein kinase with no lysine 2; protein kinase, lysine deficient 2; RGD1307284; SDCCAG43; Serine/threonine-protein kinase WNK2; serologically defined colon cancer antigen 43; WNK lysine deficient protein kinase 2; Wnk2; X83337
Common Name WNK2
Gene Symbol WNK2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GTSSSMTAESSPRSMLGYDRDGRQVASDSHVVPSVPQDVPAFVRPARVEPTDRDGGEAGESSAEPPPSDMGTVGGQASHPQTLGARALGSPRKRPEQQDVSSPAKTVGRFSVVSTQDEWTLASPHSLR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.