missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human WISP-3 Recombinant Protein, PeproTech®

Product Code. 17830123
Click to view available options
Quantity:
100 μg
2 x 250 μg
20 μg
250 μg
4 x 250 μg
5 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
Pack of 2
Pack of 4
This item is not returnable. View return policy

Product Code. 17830123

Brand: Gibco™ 120201MG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

120-20-1MG will be provided as 4 x 250 μg (120-20-250UG). 120-20-500UG will be provided as 2 x 250 μg (120-20-250UG). Recombinant Human WISP-3 is a 36.8 kDa protein containing 331 amino acid residues. It is composed of four distinct structural domains (modules): the IGF binding protein (IGFBP) domain, the von Willebrand Factor C (VWFC) domain, the thrombospondin type-1 repeat (TSP type-1) domain, and a C-terminal cysteine knot-like (CTCK) domain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

WISP3 is a member of the WNT1 inducible signaling pathway (WISP) protein subfamily, which belongs to the connective tissue growth factor (CTGF) family. WNT1 is a member of a family of cysteine-rich, glycosylated signaling proteins that mediate diverse developmental processes. The CTGF family members are characterized by four conserved cysteine-rich domains: insulin-like growth factor-binding domain, von Willebrand factor type C module, thrombospondin domain and C-terminal cystine knot-like domain. WISP3 is overexpressed in colon tumors. It may be downstream in the WNT1 signaling pathway that is relevant to malignant transformation. Mutations of the WISP3 gene are associated with progressive pseudorheumatoid dysplasia, an autosomal recessive skeletal disorder, indicating that the gene is essential for normal postnatal skeletal growth and cartilage homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95389
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 8838
Molecular Weight (g/mol) 36.8
Name Human WISP-3
Quantity 4 x 250 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias CCN family member 6; CCN6; Cellular communication network factor 6; ENSMUSG00000062074; Gm735; LIBC; PPAC; PPD; RGD1564120; UNQ462/PRO790/PRO956; Wisp3; WISP-3; WNT1 inducible signaling pathway protein 3; WNT1-inducible-signaling pathway protein 3
Gene Symbol CCN6
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Sequence TGPLDTTPEG RPGEVSDAPQ RKQFCHWPCK CPQQKPRCPP GVSLVRDGCG CCKICAKQPG EICNEADLCD PHKGLYCDYS VDRPRYETGV CAYLVAVGCE FNQVHYHNGQ VFQPNPLFSC LCVSGAIGCT PLFIPKLAGS HCSGAKGGKK SDQSNCSLEP LLQQLSTSYK TMPAYRNLPL IWKKKCLVQA TKWTPCSRTC GMGISNRVTN ENSNCEMRKE KRLCYIQPCD SNILKTIKIP KGKTCQPTFQ LSKAEKFVFS GCSSTQSYKP TFCGICLDKR CCIPNKSKMI TIQFDCPNEG SFKWKMLWIT SCVCQRNCRE PGDIFSELKI L
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.