missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human WISP-1 Recombinant Protein, PeproTech®

Product Code. 30088400
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
Pack of 2
This item is not returnable. View return policy

Product Code. 30088400

Brand: Gibco™ 1201801M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

120-18-1MG will be provided as 2 x 500 μg (120-18-500UG). Recombinant Human WISP-1 is a 38.1 kDa protein containing 345 amino acid residues. It is composed of four distinct structural domains (modules): the IGF binding protein (IGFBP) domain, the von Willebrand Factor C (VWFC) domain, the thrombospondin type-1 repeat (TSP type-1) domain, and a C-terminal cysteine knot-like (CTCK) domain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene encodes a member of the WNT1 inducible signaling pathway (WISP) protein subfamily, which belongs to the connective tissue growth factor (CTGF) family. WNT1 is a member of a family of cysteine-rich, glycosylated signaling proteins that mediate diverse developmental processes. The CTGF family members are characterized by four conserved cysteine-rich domains: insulin-like growth factor-binding domain, von Willebrand factor type C module, thrombospondin domain and C-terminal cystine knot-like domain. This gene may be downstream in the WNT1 signaling pathway that is relevant to malignant transformation. It is expressed at a high level in fibroblast cells, and overexpressed in colon tumors. The encoded protein binds to decorin and biglycan, two members of a family of small leucine-rich proteoglycans present in the extracellular matrix of connective tissue, and possibly prevents the inhibitory activity of decorin and biglycan in tumor cell proliferation. It also attenuates p53-mediated apoptosis in response to DNA damage through activation of the Akt kinase. It is 83% identical to the mouse protein at the amino acid level. Alternative splicing of this gene generates 2 transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95388
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 8840
Molecular Weight (g/mol) 38.1 kDa
Name Human WISP-1
Purification Method Purified
Quantity 2 x 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias AW146261; CCN family member 4; Ccn4; CTC-458A3.8; Elm1; ELM-1; WISP1; WISP-1; WISP1c; WISP1i; WISP1tc; WNT1 induced secreted protein 1; WNT1 inducible signaling pathway protein 1; Wnt-1 inducible signaling pathway protein 1; Wnt-1-induced secreted protein; WNT1-inducible-signaling pathway protein 1
Common Name WISP1
Gene Symbol CCN4
Biological Activity Determined by the dose-dependant proliferation of the MCF-7 cell line. The expected ED50 for this effect is 1.0-3.0 ug/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
Content And Storage -20°C
Activity Determined by the dose-dependant proliferation of the MCF-7 cell line. The expected ED50 for this effect is 1.0-3.0 ug/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.