missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human WIF-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB134-050
Additional Details : Weight : 0.00970kg
Description
WIF-1 Monoclonal specifically detects WIF-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
WIF-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
AAD25402 | |
WIF1 | |
Mouse myeloma cell line NS0-derived recombinant human WIF-1 Gly29-Trp379 Accession # AAD25402 | |
50 μg | |
Primary | |
Detects human WIF-1 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse WIF-1 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
133015 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
WIF1, WIF-1, WNT inhibitory factor 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11197 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |