Learn More
Abnova™ Human WFDC6 Full-length ORF (NP_543017.1, 1 a.a. - 86 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00140870-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. Most WFDC gene members are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the telomeric cluster. [provided by RefSeq]
Sequence: MGLSGLLPILVPFILLGDIQEPGHAEGILGKPCPKIKVECEVEEIDQCTKPRDCPENMKCCPFSRGKKCLDFRKVSLTLYHKEELESpecifications
NP_543017.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C20orf171/MGC126649/MGC126653/WAP6/dJ461P17.11 | |
WFDC6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
140870 | |
WFDC6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGLSGLLPILVPFILLGDIQEPGHAEGILGKPCPKIKVECEVEEIDQCTKPRDCPENMKCCPFSRGKKCLDFRKVSLTLYHKEELE | |
RUO | |
WFDC6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |