Learn More
Abnova™ Human WDR92 Full-length ORF (NP_612467.1, 1 a.a. - 357 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00116143-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The WD40 repeat domain is a common structural module in eukaryotes, and proteins containing WD40 domains have a diverse range of functions, including signal transduction, cell cycle regulation, RNA splicing, and transcription (Saeki et al., 2006 [PubMed 16487927]).[supplied by OMIM]
Sequence: MSAFEKPQIIAHIQKGFNYTVFDCKWVPCSAKFVTMGNFARGTGVIQLYEIQHGDLKLLREIEKAKPIKCGTFGATSLQQRYLATGDFGGNLHIWNLEAPEMPVYSVKGHKEIINAIDGIGGLGIGEGAPEIVTGSRDGTVKVWDPRQKDDPVANMEPVQGENKRDCWTVAFGNAYNQEERVVCAGYDNGDIKLFDLRNMALRWETNIKNGVCSLEFDRKDISMNKLVATSLEGKFHVFDMRTQHPTKGFASVSEKAHKSTVWQVRHLPQNRELFLTAGGAGGLHLWKYEYPIQRSKKDSEGIEMGVAGSVSLLQNVTLSTQPISSLDWSPDKRGLCVCSSFDQTVRVLIVTKLNKISpecifications
NP_612467.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ31741 | |
WDR92 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
116143 | |
WDR92 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSAFEKPQIIAHIQKGFNYTVFDCKWVPCSAKFVTMGNFARGTGVIQLYEIQHGDLKLLREIEKAKPIKCGTFGATSLQQRYLATGDFGGNLHIWNLEAPEMPVYSVKGHKEIINAIDGIGGLGIGEGAPEIVTGSRDGTVKVWDPRQKDDPVANMEPVQGENKRDCWTVAFGNAYNQEERVVCAGYDNGDIKLFDLRNMALRWETNIKNGVCSLEFDRKDISMNKLVATSLEGKFHVFDMRTQHPTKGFASVSEKAHKSTVWQVRHLPQNRELFLTAGGAGGLHLWKYEYPIQRSKKDSEGIEMGVAGSVSLLQNVTLSTQPISSLDWSPDKRGLCVCSSFDQTVRVLIVTKLNKI | |
RUO | |
WDR92 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |