missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human WDR16 Full-length ORF (AAH25392.1, 1 a.a. - 620 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00146845-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138433

  • £280.00 / 10µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

WD repeat-containing proteins, such as WDR16, play crucial roles in a wide range of physiologic functions, including signal transduction, RNA processing, remodeling the cytoskeleton, regulation of vesicular traffic, and cell division (Silva et al., 2005 [PubMed 15967112]).[supplied by OMIM]

Sequence: MDNKISPEAQVAELELDAVIGFNGHVPTGLKCHPDQEHMIYPLGCTVLIQAINTKEQNFLQGHGNNVSCLAISRSGEYIASGQVTFMGFKVDIILWDYKNRELLARLSLHKGKIEALAFSPNDLYLVSLGGPDDGSVVVWSIAKRDAICGSPAAGLNVGNATNVIFSRCRDEMFMTAGNGTIRVWELDLPNRKIWPTESQTGQLKRIVMSIGVDDDDSFFYLGTTTGDILKMNPRTKLLTDVGPAKDKFSLGVSAIRCLKMGGLLVGSGAGLLVFCKSPGYKPIKKIQLQGGITSITLRGEGHQFLVGTEESHIYRVSFTDFKETLIATCHFDAVKDIVFPFGTAELFATCAKKDIRVWHTSSNRELLRITVPNMTCHGIDFMRDGKSIISAWNDGKIRAFAPETGRLMYVINNAHRIGVTAIATTSDCKRVISGGGEGEVRVWQIGCQTQKLEEALKEHKSSVSCIRVKRNNEECVTASTDGTCIIWDLVRLRRNQMILANTLFQCVCYHPEEFQIITSGTDRKIAYWEVFDGTVIRELEGSLSGSINGMDITQEGVHFVTGGNDHLVKVWDYNEGEVTHVGVGHSGNITRIRISPGNQYIVSVSADGAILRWKYPYTS
Specifications

Specifications

AAH25392.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
94.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ37528/WDRPUH
WDR16
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
146845
WDR16 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDNKISPEAQVAELELDAVIGFNGHVPTGLKCHPDQEHMIYPLGCTVLIQAINTKEQNFLQGHGNNVSCLAISRSGEYIASGQVTFMGFKVDIILWDYKNRELLARLSLHKGKIEALAFSPNDLYLVSLGGPDDGSVVVWSIAKRDAICGSPAAGLNVGNATNVIFSRCRDEMFMTAGNGTIRVWELDLPNRKIWPTESQTGQLKRIVMSIGVDDDDSFFYLGTTTGDILKMNPRTKLLTDVGPAKDKFSLGVSAIRCLKMGGLLVGSGAGLLVFCKSPGYKPIKKIQLQGGITSITLRGEGHQFLVGTEESHIYRVSFTDFKETLIATCHFDAVKDIVFPFGTAELFATCAKKDIRVWHTSSNRELLRITVPNMTCHGIDFMRDGKSIISAWNDGKIRAFAPETGRLMYVINNAHRIGVTAIATTSDCKRVISGGGEGEVRVWQIGCQTQKLEEALKEHKSSVSCIRVKRNNEECVTASTDGTCIIWDLVRLRRNQMILANTLFQCVCYHPEEFQIITSGTDRKIAYWEVFDGTVIRELEGSLSGSINGMDITQEGVHFVTGGNDHLVKVWDYNEGEVTHVGVGHSGNITRIRISPGNQYIVSVSADGAILRWKYPYTS
RUO
WDR16
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers