missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human WDR12 (aa 1-83) Control Fragment Recombinant Protein

Product Code. 30206966
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206966

Brand: Invitrogen™ RP96107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57635 (PA5-57635. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein is highly similar to the mouse WD repeat domain 12 protein at the amino acid level. The protein encoded by this gene is a component of a nucleolar protein complex that affects maturation of the large ribosomal subunit.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9GZL7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55759
Name Human WDR12 (aa 1-83) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4933402C23Rik; fb24f09; HAMAP-Rule:MF_03029}; MNCb-5414; nuclear protein Ytm1; ribosome biogenesis protein wdr12; ribosome biogenesis protein WDR12 {ECO:0000255; WD repeat domain 12; WD repeat domain 12 protein; WD repeat-containing protein 12; WD repeat-containing protein 12 {ECO:0000255; WDR12; wdr12 protein; wu:fb24f09; YTM1; Ytm1p; zgc:55609
Common Name WDR12
Gene Symbol WDR12
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MAQLQTRFYTDNKKYAVDDVPFSIPAASEIADLSNIINKLLKDKNEFHKHVEFDFLIKGQFLRMPLDKHMEMENISSEEVVEI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.