Learn More
Abnova™ Human WDR1 Full-length ORF (NP_059830.1, 1 a.a. - 606 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009948-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein containing 9 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, mostly including a trp-asp at the C-terminal end. WD domains are involved in protein-protein interactions. The encoded protein may help induce the disassembly of actin filaments. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MPYEIKKVFASLPQVERGVSKIIGGDPKGNNFLYTNGKCVILRNIDNPALADIYTEHAHQVVVAKYAPSGFYIASGDVSGKLRIWDTTQKEHLLKYEYQPFAGKIKDIAWTEDSKRIAVVGEGREKFGAVFLWDSGSSVGEITGHNKVINSVDIKQSRPYRLATGSDDNCAAFFEGPPFKFKFTIGDHSRFVNCVRFSPDGNRFATASADGQIYIYDGKTGEKVCALGGSKAHDGGIYAISWSPDSTHLLSASGDKTSKIWDVSVNSVVSTFPMGSTVLDQQLGCLWQKDHLLSVSLSGYINYLDRNNPSKPLHVIKGHSKSIQCLTVHKNGGKSYIYSGSHDGHINYWDSETGENDSFAGKGHTNQVSRMTVDESGQLISCSMDDTVRYTSLMLRDYSGQGVVKLDVQPKCVAVGPGGYAVVVCIGQIVLLKDQRKCFSIDNPGYEPEVVAVHPGGDTVAIGGVDGNVRLYSILGTTLKDEGKLLEAKGPVTDVAYSHDGAFLAVCDASKVVTVFSVADGYSENNVFYGHHAKIVCLAWSPDNEHFASGGMDMMVYVWTLSDPETRVKIQDAHRLHHVSSLAWLDEHTLVTTSHDASVKEWTITYSpecifications
NP_059830.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
92.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AIP1/NORI-1 | |
WDR1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9948 | |
WDR1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPYEIKKVFASLPQVERGVSKIIGGDPKGNNFLYTNGKCVILRNIDNPALADIYTEHAHQVVVAKYAPSGFYIASGDVSGKLRIWDTTQKEHLLKYEYQPFAGKIKDIAWTEDSKRIAVVGEGREKFGAVFLWDSGSSVGEITGHNKVINSVDIKQSRPYRLATGSDDNCAAFFEGPPFKFKFTIGDHSRFVNCVRFSPDGNRFATASADGQIYIYDGKTGEKVCALGGSKAHDGGIYAISWSPDSTHLLSASGDKTSKIWDVSVNSVVSTFPMGSTVLDQQLGCLWQKDHLLSVSLSGYINYLDRNNPSKPLHVIKGHSKSIQCLTVHKNGGKSYIYSGSHDGHINYWDSETGENDSFAGKGHTNQVSRMTVDESGQLISCSMDDTVRYTSLMLRDYSGQGVVKLDVQPKCVAVGPGGYAVVVCIGQIVLLKDQRKCFSIDNPGYEPEVVAVHPGGDTVAIGGVDGNVRLYSILGTTLKDEGKLLEAKGPVTDVAYSHDGAFLAVCDASKVVTVFSVADGYSENNVFYGHHAKIVCLAWSPDNEHFASGGMDMMVYVWTLSDPETRVKIQDAHRLHHVSSLAWLDEHTLVTTSHDASVKEWTITY | |
RUO | |
WDR1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |