missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human WASP Antibody, R&D Systems™

Mouse Monoclonal Antibody
£170.00 - £369.00
Specifications
| Antigen | WASP |
|---|---|
| Clone | 336925 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18797539
![]() |
R&D Systems
MAB3070-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18149860
![]() |
R&D Systems
MAB3070 |
100 μg |
£369.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WASP Monoclonal specifically detects WASP in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| WASP | |
| LYOPH | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P42768 | |
| 7454 | |
| E. coli-derived recombinant human WASP Lys230-Gln310 Accession # P42768 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human WASP in direct ELISAs and Western blots. |
| 336925 | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IMD2THC, Thrombocytopenia 1, WAS, WASP, WiskTHC1, wiskthrombocytopenia 1 (X-linked) | |
| WAS | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title