missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human WASF2 Full-length ORF (NP_008921.1, 1 a.a. - 498 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010163-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122016

  • £280.00 / 10µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the Wiskott-Aldrich syndrome protein family. The gene product is a protein that forms a multiprotein complex that links receptor kinases and actin. Binding to actin occurs through a C-terminal verprolin homology domain in all family members. The multiprotein complex serves to tranduce signals that involve changes in cell shape, motility or function. The published map location (PMID:10381382) has been changed based on recent genomic sequence comparisons, which indicate that the expressed gene is located on chromosome 1, and a pseudogene may be located on chromosome X. [provided by RefSeq]

Sequence: MPLVTRNIEPRHLCRQTLPSVRSELECVTNITLANVIRQLGSLSKYAEDIFGELFTQANTFASRVSSLAERVDRLQVKVTQLDPKEEEVSLQGINTRKAFRSSTIQDQKLFDRNSLPVPVLETYNTCDTPPPLNNLTPYRDDGKEALKFYTDPSYFFDLWKEKMLQDTKDIMKEKRKHRKEKKDNPNRGNVNPRKIKTRKEEWEKMKMGQEFVESKEKLGTSGYPPTLVYQNGSIGCVENVDASSYPPPPQSDSASSPSPSFSEDNLPPPPAEFSYPVDNQRGSGLAGPKRSSVVSPSHPPPAPPLGSPPGPKPGFAPPPAPPPPPPPMIGIPPPPPPVGFGSPGTPPPPSPPSFPPHPDFAAPPPPPPPPAADYPTLPPPPLSQPTGGAPPPPPPPPPPGPPPPPFTGADGQPAIPPPLSDTTKPKSSLPAVSDARSDLLSAIRQGFQLRRVEEQREQEKRDVVGNDVATILSRRIAVEYSDSEDDSSEFDEDDWSD
Specifications

Specifications

NP_008921.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
80.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SCAR2/WAVE2/dJ393P12.2
WASF2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
10163
WASF2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPLVTRNIEPRHLCRQTLPSVRSELECVTNITLANVIRQLGSLSKYAEDIFGELFTQANTFASRVSSLAERVDRLQVKVTQLDPKEEEVSLQGINTRKAFRSSTIQDQKLFDRNSLPVPVLETYNTCDTPPPLNNLTPYRDDGKEALKFYTDPSYFFDLWKEKMLQDTKDIMKEKRKHRKEKKDNPNRGNVNPRKIKTRKEEWEKMKMGQEFVESKEKLGTSGYPPTLVYQNGSIGCVENVDASSYPPPPQSDSASSPSPSFSEDNLPPPPAEFSYPVDNQRGSGLAGPKRSSVVSPSHPPPAPPLGSPPGPKPGFAPPPAPPPPPPPMIGIPPPPPPVGFGSPGTPPPPSPPSFPPHPDFAAPPPPPPPPAADYPTLPPPPLSQPTGGAPPPPPPPPPPGPPPPPFTGADGQPAIPPPLSDTTKPKSSLPAVSDARSDLLSAIRQGFQLRRVEEQREQEKRDVVGNDVATILSRRIAVEYSDSEDDSSEFDEDDWSD
RUO
WASF2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers